siRNA Details
| Gene Symbol: WFDC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130896.2 | NP_570966.2 | 6 | 706 | ||
| NM_181510.2 | NP_852611.2 | 6 | 706 | |||
| XM_017028119.1 | XP_016883608.1 | 5 | 1655 | |||
| GenBank mRNA | AF492015.1 | - | 6 | 706 | ||
| AF492016.1 | - | 6 | 706 | |||
| AL591715.1 | - | 5 | 601 | |||
| GQ472209.1 | - | 6 | 706 | |||

WFDC8
Genomic Map