siRNA Details
| Gene Symbol: ZSWIM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080603.4 | NP_542170.3 | 2 | 365 | ||
| XM_005260610.4 | XP_005260667.1 | 2 | 499 | |||
| XM_005260611.4 | XP_005260668.1 | 2 | 582 | |||
| XM_011529100.2 | XP_011527402.1 | 2 | 456 | |||
| GenBank mRNA | AK291304.1 | - | 2 | 353 | ||
| BC001672.2 | - | 2 | 333 | 2988536 | ||
| BC080525.1 | - | 2 | 333 | 4547006 | ||

ZSWIM1
Genomic Map