siRNA Details
| Gene Symbol: KIAA2013 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138346.2 | NP_612355.1 | 1, 2 | 1227 | ||
| XM_005263531.4 | XP_005263588.1 | 1, 2 | 1220 | |||
| GenBank mRNA | AB095933.1 | - | 1, 2 | 1227 | ||
| BC035033.1 | - | 1, 2 | 1230 | 5264442 | ||
| Gene Symbol: LOC728138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC157880.1 | - | 2 | 1214 | 8862702 | |

KIAA2013
Genomic Map