siRNA Details
Gene Symbol: KIAA2013 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138346.2 | NP_612355.1 | 3 | 2258 | ||
GenBank mRNA | AL833891.1 | - | 1 | 1024 | ||
BC004501.2 | - | 2 | 871 | |||
BC035033.1 | - | 3 | 2261 | 5264442 |
