siRNA Details
| Gene Symbol: OTULIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138348.4 | NP_612357.4 | 3 | 463 | ||
| XM_011514151.2 | XP_011512453.1 | 3 | 4343 | |||
| XM_011514152.2 | XP_011512454.1 | 3 | 4343 | |||
| XM_011514154.2 | XP_011512456.1 | 3 | 4343 | |||
| XM_017010015.1 | XP_016865504.1 | 2 | 4266 | |||
| GenBank mRNA | BC007706.2 | - | 1 | 36 | 3952677 | |
| BC015392.1 | - | 3 | 389 | 4430188 | ||

OTULIN
Genomic Map