siRNA Details
| Gene Symbol: TP53I13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138349.2 | NP_612358.3 | 6 | 603 | ||
| XM_017025282.1 | XP_016880771.1 | 6 | 975 | |||
| GenBank mRNA | AB110209.1 | - | 6 | 670 | ||
| AK075341.1 | - | 6 | 634 | |||
| AK093030.1 | - | 1 | 1505 | |||
| BC001593.2 | - | 6 | 603 | 2988166 | ||

TP53I13
Genomic Map