siRNA Details
| Gene Symbol: ZNF766 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010851.2 | NP_001010851.1 | 4 | 363 | ||
| XM_011527460.1 | XP_011525762.1 | 4 | 690 | |||
| XM_017027440.1 | XP_016882929.1 | 4 | 558 | |||
| XM_017027441.1 | XP_016882930.1 | 2 | 427 | |||
| GenBank mRNA | AK024074.1 | - | 5 | 415 | ||
| AK097729.1 | - | 5 | 744 | |||
| BC109201.1 | - | 4 | 488 | |||
| BC136833.1 | - | 4 | 351 | 9020824 | ||
| BC136834.1 | - | 4 | 351 | 9020825 | ||
| BX538183.1 | - | 4 | 332 | |||
| BX648899.1 | - | 4 | 364 | |||

ZNF766
Genomic Map