siRNA Details
| Gene Symbol: THAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138350.3 | NP_612359.2 | 4, 5 | 536 | ||
| XM_005263532.3 | XP_005263589.1 | 6 | 2046 | |||
| XM_011542400.2 | XP_011540702.1 | 5 | 1620 | |||
| XM_017002761.1 | XP_016858250.1 | 4, 5 | 473 | |||
| GenBank mRNA | AK096217.1 | - | 4, 5 | 494 | ||

THAP3
Genomic Map