siRNA Details
| Gene Symbol: FAM110B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147189.2 | NP_671722.1 | 5 | 1758 | ||
| XM_005251324.2 | XP_005251381.1 | 4 | 1752 | |||
| XM_005251325.3 | XP_005251382.1 | 3 | 11915 | |||
| XM_005251326.2 | XP_005251383.1 | 3 | 11940 | |||
| XM_017013948.1 | XP_016869437.1 | 4 | 4327 | |||
| GenBank mRNA | AK026141.1 | - | 2 | 1292 | ||
| AK092221.1 | - | 5 | 1758 | |||
| BC024294.1 | - | 2 | 1270 | 5440447 | ||
| DQ431182.1 | - | 1 | 878 | |||
| U79298.1 | - | 1 | 889 | |||

FAM110B
Genomic Map