siRNA Details
| Gene Symbol: IFT20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001267774.1 | NP_001254703.1 | 5 | 670 | ||
| NM_001267775.1 | NP_001254704.1 | 5 | 469 | |||
| NM_001267776.1 | NP_001254705.1 | 4 | 433 | |||
| NM_001267777.1 | NP_001254706.1 | 5 | 469 | |||
| NM_174887.3 | NP_777547.1 | 5 | 551 | |||
| GenBank mRNA | AF070643.1 | - | 1 | 602 | ||
| AY224601.1 | - | 5 | 619 | |||
| BC002640.2 | - | 5 | 531 | 3609552 | ||
| BC038094.1 | - | 4 | 399 | 3907361 | ||
| BE619475.1 | - | 5 | 405 | |||
| BQ639919.1 | - | 5 | 419 | |||

IFT20
Genomic Map