siRNA Details
| Gene Symbol: ZNF835 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005850.2 | NP_001005850.2 | 2 | 395 | ||
| XM_005259382.2 | XP_005259439.1 | 2 | 398 | |||
| XM_005259383.3 | XP_005259440.1 | 2 | 430 | |||
| GenBank mRNA | AK299565.1 | - | 2 | 395 | ||
| DC382896.1 | - | 2 | 395 | |||

ZNF835
Genomic Map