siRNA Details
| Gene Symbol: MCU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270679.1 | NP_001257608.1 | 8 | 953 | ||
| NM_001270680.1 | NP_001257609.1 | 8 | 1293 | |||
| NM_138357.2 | NP_612366.1 | 8 | 1016 | |||
| NR_073062.1 | 10 | 1181 | ||||
| XM_017016882.1 | XP_016872371.1 | 8 | 1216 | |||
| GenBank mRNA | AK025699.1 | - | 4 | 358 | ||
| AK075439.1 | - | 8 | 1003 | |||
| AK128016.1 | - | 8 | 1293 | |||
| AK301475.1 | - | 10 | 1162 | |||
| AK315519.1 | - | 8 | 999 | |||
| BC010682.1 | - | 8 | 910 | |||
| BC034235.1 | - | 8 | 1016 | 5296557 | ||

MCU
Genomic Map