siRNA Details
| Gene Symbol: ZNF625 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145233.3 | NP_660276.2 | 4 | 1367 | ||
| NR_037801.1 | 4 | 1363 | ||||
| GenBank mRNA | BC007868.2 | - | 4 | 1346 | 4303049 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF625| Gene Symbol: ZNF625 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145233.3 | NP_660276.2 | 4 | 1367 | ||
| NR_037801.1 | 4 | 1363 | ||||
| GenBank mRNA | BC007868.2 | - | 4 | 1346 | 4303049 | |