siRNA Details
Gene Symbol: ZNF439 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152262.2 | NP_689475.1 | 3 | 575 | ||
GenBank mRNA | AK027542.1 | - | 1 | 202 | ||
AL833935.1 | - | 3 | 576 | |||
BC032857.1 | - | 3 | 447 | 5268314 |
