siRNA Details
| Gene Symbol: LYRM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181705.3 | NP_859056.2 | 4, 5 | 343 | ||
| GenBank mRNA | AK000803.1 | - | 4, 5 | 314 | ||
| BC037930.1 | - | 4, 5 | 295 | 5284728 | ||
| BC047079.1 | - | 4, 5 | 273 | 5286889 | ||
| BI601105.1 | - | 4, 5 | 319 | |||
| DA996312.1 | - | 4, 5 | 345 | |||

LYRM7
Genomic Map