siRNA Details
| Gene Symbol: CARMIL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138360.3 | NP_612369.3 | 22 | 1977 | ||
| XM_011537300.2 | XP_011535602.1 | 21 | 1636 | |||
| XM_017021751.1 | XP_016877240.1 | 18 | 1537 | |||
| GenBank mRNA | AK074167.1 | - | 9 | 724 | ||

CARMIL3
Genomic Map