siRNA Details
| Gene Symbol: ZNF598 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178167.3 | NP_835461.2 | 4, 5 | 748 | ||
| GenBank mRNA | AK024487.1 | - | 2, 3 | 1473 | ||
| BC050477.1 | - | 4, 5 | 689 | 6055142 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF598| Gene Symbol: ZNF598 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178167.3 | NP_835461.2 | 4, 5 | 748 | ||
| GenBank mRNA | AK024487.1 | - | 2, 3 | 1473 | ||
| BC050477.1 | - | 4, 5 | 689 | 6055142 | ||