siRNA Details
| Gene Symbol: HN1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144570.2 | NP_653171.1 | 3, 4 | 376 | ||
| GenBank mRNA | AK023154.1 | - | 3, 4 | 374 | ||
| AK296998.1 | - | 4, 5 | 464 | |||
| AK304025.1 | - | 3, 4 | 458 | |||
| AK309909.1 | - | 2, 3 | 248 | |||
| AK316514.1 | - | 3, 4 | 432 | |||
| AY323978.1 | - | 2, 3 | 298 | |||
| AY323979.1 | - | 3, 4 | 386 | |||
| AY598330.1 | - | 3, 4 | 340 | |||
| BC014438.1 | - | 3, 4 | 351 | 4877757 | ||
| BC060853.1 | - | 3, 4 | 370 | 5296086 | ||
| BM837125.1 | - | 3, 4 | 397 | |||

HN1L
Genomic Map