siRNA Details
| Gene Symbol: SPSB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001324081.1 | NP_001311010.1 | 6 | 766 | ||
| NM_080861.3 | NP_543137.2 | 6 | 788 | |||
| XM_005255673.4 | XP_005255730.1 | 6 | 978 | |||
| GenBank mRNA | AF403028.1 | - | 5 | 697 | ||
| AK057850.1 | - | 546 | ||||
| AK074190.1 | - | 6 | 752 | |||
| BC007588.1 | - | 6 | 734 | 3342825 | ||
| BC024244.1 | - | 6 | 752 | 4798873 | ||
| BC035065.1 | - | 6 | 745 | |||
| BC041897.1 | - | 5 | 1562 | 5298729 | ||
| BC047441.1 | - | 6 | 730 | 5303706 | ||
| BC065515.1 | - | 3 | 1031 | |||
| BI768095.1 | - | 6 | 742 | |||
| BQ930544.1 | - | 1 | 70 | |||

SPSB3
Genomic Map