siRNA Details
| Gene Symbol: CFD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317335.1 | NP_001304264.1 | 5 | 891 | ||
| NM_001928.3 | NP_001919.2 | 5 | 870 | |||
| GenBank mRNA | AK300963.1 | - | 5 | 865 | ||
| BC034529.1 | - | 4 | 786 | |||
| BC051001.1 | - | 5 | 810 | |||
| BI763909.1 | - | 246 | ||||
| M84526.1 | - | 5 | 818 | |||

CFD
Genomic Map