siRNA Details
| Gene Symbol: CREB3L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052854.3 | NP_443086.1 | 4 | 1060 | ||
| XM_006718380.3 | XP_006718443.1 | 4 | 1037 | |||
| GenBank mRNA | AB063321.1 | - | 4 | 681 | ||
| AK314072.1 | - | 4 | 1038 | |||
| BC014097.1 | - | 4 | 703 | 4546950 | ||
| BC015781.1 | - | 4 | 689 | 4856946 | ||

CREB3L1
Genomic Map