siRNA Details
| Gene Symbol: CCDC34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030771.1 | NP_110398.1 | 2 | 538 | ||
| NM_080654.2 | NP_542385.1 | 2 | 538 | |||
| GenBank mRNA | AF155108.1 | - | 2 | 1164 | ||
| AF382034.1 | - | 2 | 426 | |||
| AK308494.1 | - | 3 | 486 | |||
| AK313358.1 | - | 2 | 505 | |||
| AL555720.3 | - | 2 | 522 | |||
| AY598334.1 | - | 2 | 484 | |||
| BC008496.1 | - | 2 | 506 | 4276656 | ||
| BC029421.1 | - | 2 | 213 | |||

CCDC34
Genomic Map