siRNA Details
| Gene Symbol: ANKRD30A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052997.2 | NP_443723.2 | 34 | 4085 | ||
| XM_011519757.2 | XP_011518059.1 | 42 | 4629 | |||
| XM_017016883.1 | XP_016872372.1 | 22 | 2458 | |||
| GenBank mRNA | AF269087.1 | - | 34 | 4085 | ||

ANKRD30A
Genomic Map