siRNA Details
| Gene Symbol: TRIM47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033452.2 | NP_258411.2 | 6 | 1358 | ||
| XM_005257787.4 | XP_005257844.1 | 6 | 1405 | |||
| XM_005257788.4 | XP_005257845.1 | 6 | 764 | |||
| GenBank mRNA | AY026763.1 | - | 6 | 1359 | ||
| BC006153.1 | - | 5 | 627 | |||
| BC009225.2 | - | 3 | 217 | |||
| BC017299.2 | - | 6 | 790 | 5017710 | ||

TRIM47
Genomic Map