siRNA Details
| Gene Symbol: L3MBTL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173464.3 | NP_775735.2 | 15 | 1464 | ||
| XM_005258166.1 | XP_005258223.1 | 15 | 1423 | |||
| XM_006722364.2 | XP_006722427.1 | 15 | 1423 | |||
| XM_011525757.1 | XP_011524059.1 | 15 | 1423 | |||
| XM_011525758.1 | XP_011524060.1 | 15 | 1437 | |||
| XM_011525759.1 | XP_011524061.1 | 15 | 1444 | |||
| XM_011525760.1 | XP_011524062.1 | 15 | 1423 | |||
| XM_011525761.1 | XP_011524063.1 | 15 | 1437 | |||
| XM_011525762.1 | XP_011524064.1 | 15 | 1423 | |||
| XM_011525763.2 | XP_011524065.1 | 15 | 1423 | |||
| XM_011525765.2 | XP_011524067.1 | 15 | 1423 | |||
| XM_011525767.2 | XP_011524069.1 | 6 | 655 | |||
| XM_017026074.1 | XP_016881563.1 | 15 | 1423 | |||
| XM_017026075.1 | XP_016881564.1 | 15 | 1423 | |||
| XM_017026076.1 | XP_016881565.1 | 15 | 1444 | |||
| GenBank mRNA | AK091075.1 | - | 8 | 753 | ||
| AK093255.1 | - | 15 | 1423 | |||
| BC039316.1 | - | 15 | 1456 | 5298402 | ||

L3MBTL4
Genomic Map