siRNA Details
Gene Symbol: R3HDM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138774.3 | NP_620129.2 | 8 | 1240 | ||
GenBank mRNA | BC012775.2 | - | 8 | 1215 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: R3HDM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138774.3 | NP_620129.2 | 8 | 1240 | ||
GenBank mRNA | BC012775.2 | - | 8 | 1215 |