siRNA Details
| Gene Symbol: FAAP24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152266.4 | NP_689479.1 | 3, 4 | 384 | ||
| XM_005259393.3 | XP_005259450.1 | 3, 4 | 331 | |||
| GenBank mRNA | AK128668.1 | - | 3, 4 | 351 | ||
| BC003535.1 | - | 3, 4 | 304 | 3609326 | ||
| BC010170.2 | - | 3, 4 | 312 | 3659284 | ||
| BC020247.1 | - | 3, 4 | 332 | 4620233 | ||
| CD690094.1 | - | 3, 4 | 357 | |||
| DA006539.1 | - | 3, 4 | 359 | |||
| KU178830.1 | - | 2, 3 | 241 | |||
| KU178831.1 | - | 2, 3 | 183 | |||

FAAP24
Genomic Map