siRNA Details
| Gene Symbol: ANKRD44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195144.1 | NP_001182073.1 | 14 | 1639 | ||
| XM_005246945.1 | XP_005247002.1 | 14 | 1683 | |||
| XM_005246946.1 | XP_005247003.1 | 14 | 1629 | |||
| XM_005246947.2 | XP_005247004.1 | 14 | 1683 | |||
| XM_005246948.2 | XP_005247005.1 | 14 | 1683 | |||
| XM_006712838.3 | XP_006712901.1 | 14 | 1683 | |||
| XM_011512148.2 | XP_011510450.1 | 14 | 1683 | |||
| XM_017005252.1 | XP_016860741.1 | 14 | 1629 | |||
| GenBank mRNA | AK097086.1 | - | 14 | 1448 | ||
| AK301209.1 | - | 13 | 1560 | |||
| AL133087.1 | - | 8 | 892 | |||
| BC016985.2 | - | 14 | 1613 | 4395011 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001081433, AK086783, BC157918, BC157951, BC172102.

ANKRD44
Genomic Map