siRNA Details
| Gene Symbol: C12orf65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143905.2 | NP_001137377.1 | 3 | 532 | ||
| NM_001194995.1 | NP_001181924.1 | 3 | 825 | |||
| NM_152269.4 | NP_689482.1 | 3 | 616 | |||
| XM_005253630.4 | XP_005253687.1 | 4 | 636 | |||
| XM_011538980.2 | XP_011537282.1 | 3 | 1394 | |||
| GenBank mRNA | AF061733.1 | - | 3 | 458 | ||
| AK025908.1 | - | 3 | 1532 | |||
| AK095982.1 | - | 3 | 996 | |||
| AK126043.1 | - | 3 | 4387 | |||
| BC018145.1 | - | 3 | 417 | 3846678 | ||
| BC020885.1 | - | 3 | 1553 | 4665283 | ||
| BC062329.1 | - | 3 | 459 | 5484911 | ||
| BI850864.1 | - | 3 | 532 | |||
| BU507560.1 | - | 3 | 639 | |||

C12orf65
Genomic Map