siRNA Details
| Gene Symbol: RPS19BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194326.3 | NP_919307.1 | 3 | 331 | ||
| NR_130151.1 | 4 | 422 | ||||
| GenBank mRNA | AK025423.1 | - | 3 | 224 | ||
| AK055865.1 | - | 1 | 2209 | |||
| BC037573.1 | - | 3 | 341 | 5399759 | ||
| BC047711.1 | - | 3 | 223 | 6064917 | ||
| CB142920.1 | - | 4 | 328 | |||
| CR456443.1 | - | 3 | 261 | |||

RPS19BP1
Genomic Map