siRNA Details
| Gene Symbol: PLXNA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105543.1 | NP_001099013.1 | 3 | 1572 | ||
| NM_020911.1 | NP_065962.1 | 3 | 1572 | |||
| NM_181775.3 | NP_861440.2 | 4 | 1720 | |||
| XM_005250686.4 | XP_005250743.1 | 3 | 1970 | |||
| XM_006716171.3 | XP_006716234.1 | 4 | 2063 | |||
| XM_011516676.2 | XP_011514978.1 | 3 | 1478 | |||
| XM_017012779.1 | XP_016868268.1 | 3 | 1478 | |||
| GenBank mRNA | AB046770.3 | - | 1 | 49 | ||
| AK123428.1 | - | 3 | 1478 | |||
| AL137352.1 | - | 2 | 729 | |||
| AY358850.1 | - | 2 | 765 | |||
| BC028744.2 | - | 4 | 1720 | |||

PLXNA4
Genomic Map