siRNA Details
| Gene Symbol: ZNF765 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040185.1 | NP_001035275.1 | 4 | 1406 | ||
| GenBank mRNA | AK293890.1 | - | 6 | 1620 | ||
| AK316300.1 | - | 4 | 1400 | |||
| BC140724.1 | - | 4 | 1380 | 9021592 | ||
| BX647981.1 | - | 4 | 1398 | |||

ZNF765
Genomic Map