siRNA Details
| Gene Symbol: CENPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127181.2 | NP_001120653.1 | 4 | 860 | ||
| NM_001171182.1 | NP_001164653.1 | 3 | 471 | |||
| NM_033319.3 | NP_201576.1 | 4 | 860 | |||
| GenBank mRNA | AK055606.1 | - | 470 | |||
| BC007071.1 | - | 4 | 395 | 3682908 | ||
| BC019022.2 | - | 3 | 472 | |||
| BC033154.1 | - | 4 | 543 | 4139038 | ||
| BC066658.1 | - | 4 | 542 | 6172418 | ||

CENPL
Genomic Map