siRNA Details
| Gene Symbol: ELMSAN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001043318.2 | NP_001036783.1 | 12 | 3483 | ||
| NM_194278.3 | NP_919254.2 | 12 | 3862 | |||
| XM_005268204.1 | XP_005268261.1 | 12 | 3862 | |||
| XM_005268205.1 | XP_005268262.1 | 12 | 3418 | |||
| XM_005268206.1 | XP_005268263.1 | 13 | 3635 | |||

ELMSAN1
Genomic Map