siRNA Details
| Gene Symbol: TIMD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146726.1 | NP_001140198.1 | 2 | 136 | ||
| NM_138379.2 | NP_612388.2 | 2 | 136 | |||
| XM_011534694.2 | XP_011532996.1 | 2 | 107 | |||
| XM_017010021.1 | XP_016865510.1 | 2 | 107 | |||
| GenBank mRNA | AK301337.1 | - | 2 | 107 | ||
| AK312746.1 | - | 2 | 107 | |||
| BC008988.1 | - | 2 | 136 | 4184237 | ||
| JX049980.1 | - | 2 | 79 | |||
| KU178832.1 | - | 2 | 79 | |||
| KU178833.1 | - | 2 | 79 | |||

TIMD4
Genomic Map