siRNA Details
| Gene Symbol: COG7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153603.3 | NP_705831.1 | 4 | 691 | ||
| XM_017023870.1 | XP_016879359.1 | 4 | 734 | |||
| GenBank mRNA | AK093989.1 | - | 1 | 47 | ||
| BC000549.2 | - | 4 | 646 | 3163316 | ||
| BC037563.1 | - | 4 | 680 | 5393265 | ||
| KU178837.1 | - | 4 | 494 | |||

COG7
Genomic Map