siRNA Details
| Gene Symbol: ZNF300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172831.1 | NP_001166302.1 | 7 | 1705 | ||
| NM_001172832.1 | NP_001166303.1 | 5 | 1608 | |||
| NM_052860.2 | NP_443092.1 | 6 | 1650 | |||
| XM_011537702.2 | XP_011536004.1 | 7 | 1805 | |||
| GenBank mRNA | AF395541.1 | - | 6 | 1496 | ||
| AK026949.1 | - | 1 | 773 | |||
| AK095642.1 | - | 2 | 2397 | |||
| AK096465.1 | - | 6 | 1647 | |||
| AK300528.1 | - | 7 | 1626 | |||
| BC117248.1 | - | 6 | 1399 | 40125799 | ||
| CR933727.1 | - | 1 | 1117 | |||

ZNF300
Genomic Map