siRNA Details
| Gene Symbol: DHODH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001361.4 | NP_001352.2 | 3 | 407 | ||
| XM_005255827.3 | XP_005255884.1 | 3 | 712 | |||
| XM_005255829.3 | XP_005255886.1 | 1 | 174 | |||
| XM_017022990.1 | XP_016878479.1 | 3 | 487 | |||
| GenBank mRNA | AK292293.1 | - | 4 | 563 | ||
| AU130529.1 | - | 4 | 407 | |||
| BC065245.1 | - | 3 | 387 | 6064723 | ||
| BI253747.1 | - | 3 | 391 | |||
| M94065.1 | - | 4 | 389 | |||

DHODH
Genomic Map