siRNA Details
| Gene Symbol: DHODH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001361.4 | NP_001352.2 | 7 | 954 | ||
| XM_005255827.3 | XP_005255884.1 | 7 | 1259 | |||
| XM_005255829.3 | XP_005255886.1 | 6 | 896 | |||
| XM_017022990.1 | XP_016878479.1 | 7 | 1034 | |||
| GenBank mRNA | AK097300.1 | - | 2 | 1598 | ||
| AK292293.1 | - | 8 | 1110 | |||
| BC065245.1 | - | 7 | 934 | 6064723 | ||
| M94065.1 | - | 8 | 936 | |||

DHODH
Genomic Map