siRNA Details
| Gene Symbol: TMEM88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001319941.1 | NP_001306870.1 | 2 | 472 | ||
| NM_203411.1 | NP_981956.1 | 2 | 798 | |||
| XM_005256856.3 | XP_005256913.1 | 2 | 1020 | |||
| GenBank mRNA | AK000032.1 | - | 2 | 799 | ||
| BC057812.1 | - | 2 | 798 | 30332216 | ||

TMEM88
Genomic Map