siRNA Details
| Gene Symbol: MTG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138384.2 | NP_612393.2 | 9 | 832 | ||
| GenBank mRNA | AK074976.1 | - | 8 | 1311 | ||
| AK095872.1 | - | 3 | 1701 | |||
| AK125012.1 | - | 9 | 1677 | |||
| AK292515.1 | - | 9 | 674 | |||
| AK293988.1 | - | 7 | 587 | |||
| BC000920.2 | - | 7 | 541 | |||
| BC004409.1 | - | 9 | 791 | |||
| BC026039.1 | - | 9 | 808 | 5444745 | ||
| BC026725.1 | - | 9 | 711 | |||
| BC035721.1 | - | 9 | 736 | 5588933 | ||

MTG1
Genomic Map