siRNA Details
Gene Symbol: UBTD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152277.2 | NP_689490.2 | 1, 2 | 202 | ||
GenBank mRNA | BC019910.1 | - | 1, 2 | 50 | 4402070 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: UBTD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152277.2 | NP_689490.2 | 1, 2 | 202 | ||
GenBank mRNA | BC019910.1 | - | 1, 2 | 50 | 4402070 |