siRNA Details
| Gene Symbol: DAPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017920.2 | NP_001017920.2 | 2 | 191 | ||
| GenBank mRNA | AF086541.1 | - | 2 | 179 | ||
| AY324399.1 | - | 2 | 191 | |||
| BC127682.1 | - | 2 | 176 | 40127452 | ||
| BC127683.1 | - | 2 | 176 | 40127454 | ||
| BC140719.1 | - | 2 | 178 | 9021587 | ||
| HQ179934.1 | - | 2 | 150 | |||
| HQ179935.1 | - | 2 | 150 | |||
| HQ179936.1 | - | 2 | 150 | |||
| HQ179937.1 | - | 2 | 150 | |||
| HQ179938.1 | - | 2 | 150 | |||
| HQ179939.1 | - | 2 | 150 | |||

DAPL1
Genomic Map