siRNA Details
| Gene Symbol: DUSP27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080426.1 | NP_001073895.1 | 3 | 424 | ||
| XM_005245603.3 | XP_005245660.1 | 4 | 587 | |||
| XM_011510146.2 | XP_011508448.1 | 3 | 461 | |||
| GenBank mRNA | BC115387.1 | - | 4 | 544 | ||

DUSP27
Genomic Map