siRNA Details
| Gene Symbol: TMEM129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127266.1 | NP_001120738.1 | 2 | 848 | ||
| NM_138385.3 | NP_612394.1 | 2 | 848 | |||
| XM_005248038.4 | XP_005248095.1 | 2 | 720 | |||
| XM_005248039.4 | XP_005248096.1 | 2 | 289 | |||
| XM_011513594.2 | XP_011511896.1 | 2 | 298 | |||
| XM_017008834.1 | XP_016864323.1 | 2 | 585 | |||
| GenBank mRNA | AK303880.1 | - | 2 | 161 | ||
| BC009331.2 | - | 2 | 848 | |||
| BC018921.2 | - | 2 | 848 | |||
| BC126370.1 | - | 3 | 738 | 8992086 | ||

TMEM129
Genomic Map