siRNA Details
| Gene Symbol: MEX3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001093725.1 | NP_001087194.1 | 2 | 1784 | ||
| GenBank mRNA | AK024097.1 | - | 1 | 744 | ||
| AK095491.1 | - | 1 | 644 | |||
| AK125482.1 | - | 1 | 1286 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MEX3A| Gene Symbol: MEX3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001093725.1 | NP_001087194.1 | 2 | 1784 | ||
| GenBank mRNA | AK024097.1 | - | 1 | 744 | ||
| AK095491.1 | - | 1 | 644 | |||
| AK125482.1 | - | 1 | 1286 | |||