siRNA Details
| Gene Symbol: MEX3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001093725.1 | NP_001087194.1 | 2 | 1804 | ||
| GenBank mRNA | AK024097.1 | - | 1 | 764 | ||
| AK095491.1 | - | 1 | 664 | |||
| AK125482.1 | - | 1 | 1306 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001029890.

MEX3A
Genomic Map