siRNA Details
| Gene Symbol: CHMP4C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152284.3 | NP_689497.1 | 2, 3 | 548 | ||
| GenBank mRNA | AB120734.1 | - | 2, 3 | 380 | ||
| AK000049.1 | - | 2, 3 | 510 | |||
| AK314873.1 | - | 2, 3 | 547 | |||
| AY329086.1 | - | 2, 3 | 382 | |||
| BC014321.1 | - | 2, 3 | 552 | 3828410 | ||
| BX492886.1 | - | 2, 3 | 568 | |||

CHMP4C
Genomic Map