siRNA Details
| Gene Symbol: BICDL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207311.2 | NP_997194.2 | 6 | 1155 | ||
| XM_005253991.4 | XP_005254048.1 | 6 | 1180 | |||
| XM_006719694.3 | XP_006719757.1 | 7 | 1194 | |||
| XM_006719696.3 | XP_006719759.1 | 7 | 1193 | |||
| XM_011538999.2 | XP_011537301.1 | 7 | 1196 | |||
| XM_011539000.2 | XP_011537302.1 | 6 | 612 | |||
| GenBank mRNA | AK129960.1 | - | 7 | 1098 | ||
| AK301580.1 | - | 2 | 140 | |||

BICDL1
Genomic Map