siRNA Details
| Gene Symbol: TMEM169 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142310.1 | NP_001135782.1 | 4 | 900 | ||
| NM_001142311.1 | NP_001135783.1 | 3 | 814 | |||
| NM_001142312.1 | NP_001135784.1 | 3 | 808 | |||
| NM_138390.3 | NP_612399.1 | 3 | 810 | |||
| XM_006712851.1 | XP_006712914.1 | 3 | 1014 | |||
| XM_011512173.2 | XP_011510475.1 | 3 | 1205 | |||
| GenBank mRNA | AK091582.1 | - | 3 | 757 | ||
| BC008604.1 | - | 3 | 766 | 4180869 | ||
| CR933667.1 | - | 3 | 859 | |||

TMEM169
Genomic Map